Learn More
CD31/PECAM-1 Antibody (TLD-3A12), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-63701
Product Code. 18033224
  • ONLINE EXCLUSIVE PRICE
    € 565.00
    € 534.45 / 0.10mg
    Save € 30.55
    5% Off
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Human, Mouse, Rat, Rabbit samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD31/PECAM-1
Monoclonal
0.1 mg/mL
Flow Cytometry 1:10-1:1000
PECAM1
This CD31/PECAM-1 Antibody (TLD-3A12) was developed against activated, Lewis rat derived microglial cells .
Protein G purified
RUO
Primary
Recognizes rat PECAM-1 (CD31), a 661 amino acid type 1 transmembrane protein expressed primarily on endothelial cells, platelets and leucocytes.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Flow Cytometry
TLD-3A12
Biotin
adhesion molecule, CD31, CD31 antigen, CD31/EndoCAM, EndoCAM, FLJ34100, FLJ58394, GPIIA′, PECA1, PECAM-1, PECAM-1, CD31/EndoCAM, platelet endothelial cell adhesion molecule, platelet endothelial cell adhesion molecule-1, platelet/endothelial cell adhesion molecule
Mouse
82.5 kDa
0.1 mg
Mesenchymal Stem Cell Markers, Stem Cell Markers
5175
Mouse, Rat