Learn More
CD314 Mouse anti-Human, APC, Clone: 1D11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech APC-65188-100TESTS
This item is not returnable.
View return policy
Description
The CD314 antibody from Proteintech is a mouse monoclonal antibody generated with nkl cell line of human CD314. This antibody recognizes human antigen. The CD314 antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
| CD314 | |
| Monoclonal | |
| 10 μL/Test | |
| Mouse | |
| Primary | |
| Human |
| Flow Cytometry | |
| 1D11 | |
| APC | |
| 100 Tests | |
| 22914 | |
| IgG1 κ |