Learn More
CD314 (NKG2D) Mouse anti-Human, BV750, Clone: 1D11, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747025
This item is not returnable.
View return policy
Description
CD314 (NKG2D) Monoclonal antibody specifically detects CD314 (NKG2D) in Human samples. It is validated for Flow Cytometry
Specifications
| CD314 (NKG2D) | |
| Monoclonal | |
| Brilliant Violet 750 | |
| CD314; KLRK1; KLR; NKG2D; NKG2-D; NK cell receptor D | |
| NKL cells | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 1D11 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |