Learn More
CD32 Mouse anti-Human, Brilliant Violet 711, Clone: 3D3, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744257
Description
The 3D3 monoclonal antibody specifically recognizes FcγRII (CD32), a 40 kDa, polymorphic type I transmembrane glycoprotein that serves as a low affinity receptor for aggregated IgG. This highly glycosylated molecule (encoded by at least two different genes) is expressed on monocytes, granulocytes, platelets and B cells. Unlike the FLI28.26 mAb, the 3D3 mAb detected a polymorphic CD32 antigen expressed on B cells of all donors, but only on platelets, monocytes and granulocytes of some donors. The platelets from 3D3+ donors respond to certain stimulatory mAb such as CD165 (clone SN2) which results in aggregation. On the other hand, the platelets from 3D3 negative donors do not form aggregates after stimulation. Individuals can be divided into two groups as responder and non-responder depending on expression, or non-expression, of 3D3. In comparison to the 3D3 mAb, the FLI8.26 mAb detects a monomorphic CD32 antigen expressed on all human donors.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
| CD32 | |
| 3D3 | |
| Brilliant Violet 711 | |
| FcγRII, CD32a/FcγRIIa/FCGR2A, CD32b/FcγRIIb/FCGR2B | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.