missing translation for 'onlineSavingsMsg'
Learn More

CD32 Rat, Clone: 2.4G2, enQuireBio™

Rat Monoclonal Antibody

€ 156.45 - € 526.05

Specifications

Antigen CD32
Clone 2.4G2
Concentration 0.5mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15982578
View Documents
EnquireBio
QAB87-100ug
100 μg
€ 156.45
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15902588
View Documents
EnquireBio
QAB87-500ug
500 μg
€ 288.75
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15992578
View Documents
EnquireBio
QAB87-1mg
1 mg
€ 526.05
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

CD32
0.5mg/mL
Monoclonal
Rat
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
14131
IgG2b
Protein A/G Purified
2.4G2
Flow Cytometry
Unconjugated
RUO
XP_006496722
FCGR2b
Primary
Store at 2-4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.