missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP4C Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | PPP4C Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMAEISDL DRQIEQLRRC ELIKESEVKA LCAKAREILV EESNVQRVDS PVTVCGDIHG QFYDLKELFR VGGDVPETNY LFMGDFVDRG FYSVETFLLL LALKVRYPDR ITLIRGNHES RQITQVYGFY DECLRKYGSV TVWRYCTEIF DYLSLSAIID GKIFCVHGGL SPSIQTLDQI RTIDRKQEVP HDGPMCDLLW SDPEDTTGWG VSPRGAGYLF GSDVVAQFNA ANDIDMICRA HQLVMEGYKW HFNETVLTVW SAPNYCYRCG NVAAILELDE HLQKDFIIFE AAPQETRGIP SKKPVADYFL |
| Buffer | The PPP4C solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction