Learn More
CD321 (JAM-1) Mouse anti-Human, Brilliant Violet 650, Clone: M.Ab.F11, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744066
Description
The M.Ab.F11 monoclonal antibody specifically binds to CD321 which is also known as JAM-1 (Junctional adhesion molecule 1), Junctional adhesion molecule A (JAM-A), and F11 Receptor (F11R). CD321 is a 32-35 kDa type I transmembrane glycoprotein that includes two extracellular immunoglobulin-like domains. CD321 is expressed on platelets, leucocytes, red blood cells, endothelial cells, epithelial cells, and various cell lines. CD321 functions as an adhesion receptor molecule on platelets. It also supports the tight junction formation between endothelial cells, where it may regulate the transendothelial migration of leucocytes, and epithelial cells. M.Ab.F11 is a stimulatory antibody that can induce morphological changes, granule secretion, and aggregation in human platelets.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
| CD321 (JAM-1) | |
| M.Ab.F11 | |
| Brilliant Violet 650 | |
| JAM-1, Platelet F11 receptor, F11R, JAM-A, PAM-1 | |
| Human Platelet Membranes | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.