Learn More
CD33 (Human) Matched Antibody Pair, Abnova™
Matched Antibody Pair
Brand:  Abnova H00000945-AP21.1Set
Product Code. 16073788
  • € 1580.00 / 1 set
Specifications
CD33
Unconjugated
FLJ00391/SIGLEC-3/SIGLEC3/p67
1 Set
Primary
Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-CD33 (100 ug) 2. Detection antibody: mouse purified polyclonal anti-CD33 (20 ug) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
ELISA
CD33 molecule
CD33
RUO
945
Human
Ab Pair