Learn More
CD33 Monoclonal Antibody (OTI1E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA506319S
Encompass_Preferred
 View more versions of this product
Product Code. 17555742
  • € 242.00 / 30µL
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
CD33
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NYG5
Mouse
Affinity chromatography
RUO
51529
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Western Blot
OTI1E10
Unconjugated
ANAPC11
CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3
Full length human recombinant protein of human ANAPC11 produced in HEK293T cell.
30 μL