missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD33 Monoclonal Antibody (OTI1H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506237S
This item is not returnable.
View return policy
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Western BlotSpecifications
| CD33 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P15391 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 930 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1H8 | |
| Unconjugated | |
| Cd19 | |
| CD33; CD33 antigen; CD33 antigen (gp67); CD33 molecule; CD33 molecule transcript; FLJ00391; gp67; Myeloid cell surface antigen CD33; p67; sialic acid binding Ig-like lectin 3; sialic acid-binding Ig-like lectin 3; SIGLEC3; Siglec-3 | |
| Full length human recombinant protein of human CD19 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction