Learn More
CD34 Antibody (700011), DyLight 488, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB6518K
Product Code. 18543166
  • € 680.00 / 0.10mL
Description
CD34 Monoclonal antibody specifically detects CD34 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD34
Monoclonal
DyLight 488
50mM Sodium Borate
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, CyTOF
700011
Flow Cytometry, CyTOF-ready
CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule
Mouse myeloma cell line NS0-derived recombinant mouse CD34, Thr35-Thr287, Accession # Q64314
0.1 mL
Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors
947
Store at 4°C in the dark.
IgG2a