missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (QBEnd/10) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 493.00 - € 722.00
Specifications
| Antigen | CD34 |
|---|---|
| Clone | QBEnd/10 |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.25-0.5ug/mL, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/mL, Immunohistochemistry-Frozen 0.5-1ug/mLimmunohistochemistry-Paraffin 0.5-1ug/mL, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18424252
|
Novus Biologicals
NBP2-34713-0.2mg |
0.2 mg |
€ 722.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18408591
|
Novus Biologicals
NBP2-34713-0.1mg |
0.1 mg |
€ 493.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD34 Monoclonal antibody specifically detects CD34 in Human, Primate, Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOFSpecifications
| CD34 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| PBS | |
| 947 | |
| IgG1 κ | |
| Protein A purified |
| QBEnd/10 | |
| Western Blot 0.25-0.5ug/mL, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Paraffin 0.5-1ug/mL, Immunohistochemistry-Frozen 0.5-1ug/mLimmunohistochemistry-Paraffin 0.5-1ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate, Bovine (Negative), Canine (Negative), Rat (Negative), Sheep (Negative) | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| Detergent solubilized vesicular suspension prepared from human term placenta | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title