missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Antibody (QBEnd/10 + HPCA1/763), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47909H
This item is not returnable.
View return policy
Description
CD34 Monoclonal specifically detects CD34 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Proximity Ligation Assay.
Specifications
| CD34 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| CD34 | |
| Detergent solubilized vesicular suspension prepared from human term placenta (QBEnd/10); Recombinant full-length human CD34 protein (CD34/763) (Uniprot: P28906) | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, B Cell Development and Differentiation Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Hypoxia, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neuronal Cell Markers, Neuroscience, Stem Cell Markers, Tumor Suppressors | |
| This monoclonal antibody recognizes a single chain, transmembrane, heavily glycosylated protein of 90-120kDa, which is identified as CD34. Its expression is a hallmark for identifying pluripotent hematopoietic stem or progenitor cells. Its expression is gradually lost as lineage committed progenitors differentiate. CD34 is a marker of choice for staining blasts in acute myeloid leukemia. In addition, CD34 is expressed by soft tissue tumors, such as solitary fibrous tumor and gastrointestinal stromal tumor. Its expression is also found in vascular endothelium. Additionally, it appears that proliferating endothelial cells express this molecule more than the non-proliferating endothelial cells. Anti-CD34 labels 85% of angiosarcoma and Kaposis sarcoma, but with a lower specificity. | |
| Store at 4°C in the dark. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| QBEnd/10 + HPCA1/763 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CD34 antigenhematopoietic progenitor cell antigen CD34, CD34 molecule | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction