missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34, Mouse anti-Human, Clone: 4H5E7 + 9B10D4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15264.100uL
This item is not returnable.
View return policy
Specifications
| CD34 | |
| Monoclonal | |
| Unconjugated | |
| In ascitic fluid (0.03% sodium azide). | |
| - | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 4H5E7 + 9B10D4 | |
| ELISA (1:10000) Immunofluorescence (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:1000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. | |
| CD34 molecule | |
| CD34 | |
| Recombinant protein corresponding to human CD34. | |
| RUO | |
| 947 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction