missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies CD34+ Progenitor Reprogramming Kit
Description
- The CD34+ Progenitor Reprogramming Kit contains an integrated set of tools and reagents to enrich, expand and reprogram CD34+ progenitor cells from human peripheral blood samples
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells, Pluripotent Stem Cells |
| For Use With (Application) | Cell Culture, Reprogramming |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?