missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (594708), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5748CL1
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CD35 | |
| Monoclonal | |
| CoraFluor 1 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Mouse myeloma cell line NS0-derived recombinant human CD35, Gln42-Asp1971 (His1208Arg), Accession # CAA68755 | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 594708 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction