Learn More
CD35 Antibody (7G6), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52667PE
Product Code. 18652758
  • ONLINE EXCLUSIVE PRICE
    € 630.00
    € 595.35 / 0.10mL
    Save € 34.65
    5% Off
Description
CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Functional Assay, CyTOF, Neutralization
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD35
Monoclonal
PE
complement component (3b/4b) receptor 1 (Knops blood group), KN
Purified Mouse CR1.
0.1 mL
Immunology
1378
Store at 4°C in the dark.
IgG κ
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
7G6
PBS
Rabbit
Protein A purified