missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (8C12), PE - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-52668PE
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF
Specifications
| CD35 | |
| Monoclonal | |
| PE | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Purified murine CR1 protein. | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| 8C12 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction