missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (CR1/4382R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08296AF405
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD35 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Synthetic peptide corresponding to CD35 residues within aa1939-2039 of CD35 was used as an immunogen. (Uniprot: P17927 ) | |
| 100 μg | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| CR1/4382R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction