missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (E11), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-33080G
This item is not returnable.
View return policy
Description
CD35 Monoclonal specifically detects CD35 in Human, Monkey, Baboon, Cynomolgus Monkey, Rhesus Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| CD35 | |
| Monoclonal | |
| DyLight 488 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Mouse | |
| 215 kDa | |
| 0.1 mL | |
| Primary | |
| Recognizes a protein of 210-220kDa, which is identified as the complement receptor 1 (CR1)/CD35. This monoclonal antibody is specific for a site in CR1 that is distal from the C3b-binding site, so that it is unable to block CR1 activity. This monoclonal antibody is highly specific to CR1 and shows no cross-reaction with CR2. The primary function of CR1 is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. The Knops blood group system is a system of antigens located on this protein. Follicular dendritic cells (FDC) are restricted to the B-cell regions of secondary lymphoid follicles. They are CD21+/CD35+/CD1a-. This monoclonal antibody labels follicular dendritic cells and follicular dendritic cell sarcoma. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| ELISA | |
| E11 | |
| ELISA | |
| CR1 | |
| Intact human monocytes were used as immunogen to generate the CD35 antibody (Hogg et al, 1984). | |
| Protein A or G purified | |
| RUO | |
| 1378 | |
| Human, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction