missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (rCR1/8600) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24206JF585
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| CD35 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Recombinant fragment (around aa600-900) of human CD35 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rCR1/8600 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction