missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (SPM554), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34796JF549
This item is not returnable.
View return policy
Description
CD35 Monoclonal antibody specifically detects CD35 in Human, Baboon, Cynomolgus Monkey, Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| CD35 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Intact human monocytes | |
| 0.1 mL | |
| Immunology | |
| 1378 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| SPM554 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Baboon, Cynomolgus Monkey, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction