missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD35 Antibody (To5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50177PCP
This item is not returnable.
View return policy
Description
CD35 Monoclonal specifically detects CD35 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD35 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| CR1 | |
| C3b receptor-enriched precipitates from tonsil cell membranes | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| To5 | |
| Flow Cytometry | |
| complement component (3b/4b) receptor 1 (Knops blood group), KN | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 1378 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction