missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD35 Monoclonal Antibody (E11), PE
Mouse Monoclonal Antibody
Brand: Invitrogen™ A15456
This item is not returnable.
View return policy
Description
CD35 Monoclonal Antibody for Flow
CD35 (CR1, Complement Receptor 1) is a cell membrane-bound, monomeric glycoprotein and its primary function is to serve as the cellular receptor for C3b and C4b, the most important components of the complement system leading to clearance of foreign macromolecules. CD35 protein mediates cellular binding to particles and immune complexes that have activated complement. The CD35 complex is four different allotypes (C,A,B,D) C is 160 kDa; A is 190 kDa; B is 220 kDa and D is 250 kDa. CD35 is involved in the processing of immune complexes, promoting of binding and phagocytosis of C3b complexes and inhibition of complement activation. CD35 is on neutrophils, eosinophils, monocytes, B-cells, some NK-cells, erythrocytes, myeloid leukemias, follicular dendritic reticulum cells and is negative on basophils. Decreases in expression of CD35 protein and/or mutations in its gene have been associated with gallbladder carcinomas, mesangiocapillary glomerulonephritis, systemic lupus erythematosus and sarcoidosis. Mutations in the CD35 gene have also been associated with a reduction in Plasmodium falciparum rosetting, conferring protection against severe malaria. Alternate allele-specific splice variants encoding different isoforms of CD35 have been characterized. An additional secreted form of CD35 have also been described but has not been fully characterized.
Specifications
| CD35 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with <0.1% sodium azide; pH 7.5 | |
| P17927 | |
| CR1 | |
| Human CD35 from Acute monocytic leukemia cells and normal blood monocytes. | |
| 100 μg | |
| Primary | |
| Human, Baboon, Rhesus Macaque, Chimpanzee, Cynomolgus Monkey | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry | |
| E11 | |
| PE | |
| CR1 | |
| C3 binding protein; C3b/C4b receptor; C3-binding protein; C3BR; C4BR; CD35; CD35 antigen; complement component (3b/4b) receptor 1 (Knops blood group); Complement Component (3b/4b) receptor 1 including Knops blood group system; complement component 3b/4b receptor 1 (Knops blood group); complement receptor type 1; CR1; KN; Knops blood group antigen | |
| Mouse | |
| purified | |
| RUO | |
| 102117401, 1378, 449643, 718120 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction