missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody (877302), Alexa Fluor 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB19553AF405
This item is not returnable.
View return policy
Description
CD36 Monoclonal specifically detects CD36 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| CD36/SR-B3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD36 | |
| Partial recombinant human CD36 protein (amino acids 30-439) [UniProt P16671] | |
| 100 μL | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Store at 4C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| 877302 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction