missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-145AF594
This item is not returnable.
View return policy
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoblotting.
Specifications
| CD36/SR-B3 | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoblotting | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Rabbit | |
| 0.1 mL | |
| 948 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoblot | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CD36 | |
| A synthetic peptide mapping to a region of human CD36 between residues 300-400. [UniProtNo. P16671] | |
| Primary | |
| Human, Mouse, Amphibian | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction