missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-145H
This item is not returnable.
View return policy
Description
CD36 Polyclonal specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunoblotting.
Specifications
| CD36/SR-B3 | |
| Polyclonal | |
| Western Blot, Simple Western, Immunoblotting | |
| P16671 | |
| CD36 | |
| A synthetic peptide mapping to a region of human CD36 between residues 300-400. [UniProt# P16671] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human.Mouse reactivity reported in scientific literature (PMID: 24163073) Immunogen sequence has 95% homology to rat, monkey, Golden hamster and rabbit, 90% homology to mouse, and 75% homology to bovine. | |
| Store at 4C in the dark. |
| Western Blot, Immunoblot | |
| HRP | |
| PBS with No Preservative | |
| CD36 antigen, CD36 molecule (thrombospondin receptor), FA6-152, FATCHDS7, Fatty acid translocase, Glycoprotein IIIb, GP3Bthrombospondin receptor), GPIIIB, Leukocyte differentiation antigen CD36, PAS IV, PAS-4, Platelet collagen receptor, platelet glycoprotein 4, Platelet glycoprotein IV, scavenger receptor class B, member 3, Thrombospondin receptor | |
| Rabbit | |
| 75 kDa | |
| 0.1 mL | |
| Cancer, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Human, Mouse, Amphibian | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction