missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB400-145PE
This item is not returnable.
View return policy
Description
CD36 Polyclonal antibody specifically detects CD36 in Human, Mouse, Amphibian samples. It is validated for Flow Cytometry
Specifications
| CD36 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Amphibian | |
| Purified |
| Flow Cytometry | |
| PE | |
| BDPLT10, CD36 antigen (collagen type I receptor, thrombospondin receptor), CD36 molecule (thrombospondin receptor), CHDS7, cluster determinant 36, FAT, Fatty acid translocase, Glycoprotein IIIb, GP3B, GP4, GPIIIB, GPIV, leukocyte differentiation antigen CD36, PAS IV, PAS-4 protein, PASIV, platelet glycoprotein 4, platelet glycoprotein IV, SCARB3, scavenger receptor class B, member 3 | |
| This CD36 Antibody was developed against a synthetic peptide mapping to a region of human CD36 between residues 300-400 [Uniprot# P16671] | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Cellular Markers, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lipid and Metabolism, Stem Cell Markers | |
| 948 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction