missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD36 Mouse anti-Human, Brilliant Violet 786, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 745554
This item is not returnable.
View return policy
Description
CD36 Monoclonal antibody specifically detects CD36 in Human samples. It is validated for Flow cytometry
Specifications
| CD36 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| CLB-IVC7 | |
| Brilliant Violet 786 | |
| GPIIIb; GPIV; OKM-5; PASIV; FAT; SCARB3 | |
| Human Monocytes | |
| RUO | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon Brilliant Violet 786 under optimal conditions that minimize unconjugated dye and antibody. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction