missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD366 (TIM-3) Mouse anti-Mouse, BUV395, Clone: 5D12/TIM-3 (also known as 5D12), BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747620
This item is not returnable.
View return policy
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD366 (TIM-3) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 5D12/TIM-3 (also known as 5D12) | |
| Brilliant Ultraviolet 395 | |
| 50 μg | |
| Primary | |
| Refrigerate (2 to 8°C) | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction