Learn More
CD366 (TIM-3) Rat anti-Human, Brilliant Violet 421, Clone: 344823, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 747962
This item is not returnable.
View return policy
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Human samples. It is validated for Flow Cytometry.
Specifications
| CD366 (TIM-3) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| CD366 (TIM-3) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 344823 | |
| Brilliant Violet 421 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2a κ |