Learn More
CD366 (TIM-3) Rat anti-Human, Brilliant Violet 421, Clone: 344823, BD OptiBuild™
Rat Monoclonal Antibody
Brand:  BD Biosciences 747962
Product Code. 17229604
  • € 510.00 / 50µg
Description
CD366 (TIM-3) Monoclonal antibody specifically detects CD366 (TIM-3) in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD366 (TIM-3)
Monoclonal
0.2 mg/mL
Rat
RUO
CD366 (TIM-3)
Refrigerate (2 to 8°C)
Flow Cytometry
344823
Brilliant Violet 421
50 μg
Primary
Human
IgG2a κ