Learn More
Invitrogen™ CD367 (Clec4A, DCIR) Monoclonal Antibody (9E8), PE, eBioscience™

Mouse Monoclonal Antibody
Brand: Invitrogen™ 12-9875-42
Description
Description: This 9E8 monoclonal antibody reacts with human CD367, also known as dendritic cell immunoreceptor (DCIR) or Clec4A. This C-type lectin is expressed on monocytes, granulocytes, B cells, and plasmacytoid dendritic cells in peripheral blood. DCIR is also expressed on mature and immature monocyte-derived dendritic cells in vitro. DCIR is not expressed on NK or T cells. With its cytoplasmic ITIM domain, DCIR is an inhibitory receptor that recruits the SHP-1 and SHP-2 phosphatases upon ligand binding. Studies have demonstrated that DCIR plays a role in the interaction between dendritic cells and HIV-1. Applications Reported: This 9E8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9E8 antibody has been pre-titrated and tested by flow cytometric analysis of human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in inflammatory and immune response. Multiple transcript variants encoding distinct isoforms have been identified for this gene. This gene is closely linked to other CTL/CTLD superfamily members on chromosome 12p13 in the natural killer gene complex region.
Specifications
| CD367 (Clec4A, DCIR) | |
| Monoclonal | |
| 5 μL/Test | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Q9UMR7 | |
| CLEC4A | |
| Affinity chromatography | |
| RUO | |
| 50856 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 9E8 | |
| PE | |
| CLEC4A | |
| CD367; CLEC4A; Clec4a2; Clecsf6; C-type (calcium dependent, carbohydrate recognition domain) lectin, superfamily member 6; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 6; C-type lectin DDB27; C-type lectin domain family 4 member A; C-type lectin domain family 4, member A; C-type lectin domain family 4, member a2; C-type lectin superfamily member 6; DCIR; Dcir1; Dcir2; DDB27; Dendritic cell immunoreceptor; dendritic cell inhibitory receptor 1; dendritic cell inhibitory receptor 2; HDCGC13P; immunoreceptor; lectin-like immunoreceptor; LLIR | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.