missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Antibody (G28-1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 250.00 - € 666.75
Specifications
| Antigen | CD37 |
|---|---|
| Clone | G28-1 |
| Concentration | 1 mg/mL |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18659808
|
Novus Biologicals
NBP3-09051-0.025mg |
0.025 mg |
€ 250.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648427
|
Novus Biologicals
NBP3-09051-0.2mg |
0.2 mg |
€ 705.00 € 666.75 / 0.20mg Save € 38.25 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| CD37 | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
| This antibody was raised by immunizing mice with CD37. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| G28-1 | |
| Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Immunology | |
| PBS | |
| 951 | |
| IgG1 κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title