missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Antibody (IPO-24), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11462AF750
This item is not returnable.
View return policy
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Flow Cytometry, Immunofluorescence
Specifications
| CD37 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence | |
| IPO-24 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
| Spleen cells of a patient with hairy cell leukemia | |
| 0.1 mL | |
| Cellular Markers, Immunology | |
| 951 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction