missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Antibody (MB-1), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11968CL1
This item is not returnable.
View return policy
Description
CD37 Monoclonal antibody specifically detects CD37 in Human samples. It is validated for Flow Cytometry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| CD37 | |
| Monoclonal | |
| CoraFluor 1 | |
| CD37 antigentetraspanin-26, CD37 molecule, cell differentiation antigen 37, GP52-40, leukocyte antigen CD37, MGC120234, Tetraspanin-26, Tspan-26, TSPAN26leukocyte surface antigen CD37 | |
| BALB/c mice were immunized with lymph node cells from a patient with nodular poorly differentiated lymphocytic lymphoma. Immunophenotyping studies of these cells identified them as B cells expressing SIgG/lambda, as well as HLA-DR. Leu-12, and B1. | |
| 0.1 mL | |
| Cellular Markers, Immunology | |
| 951 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| MB-1 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction