missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD37 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21044-1-AP-20UL
This item is not returnable.
View return policy
Description
CD37 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It may play a role in T-cell-B-cell interactions. Alternate splicing results in multiple transcript variants encoding different isoforms.Specifications
| CD37 | |
| Polyclonal | |
| Unconjugated | |
| CD37 | |
| CD37, CD37 molecule, GP52 40, Leukocyte antigen CD37, Tetraspanin 26, Tspan 26, TSPAN26 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 12493, 29185, 951 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11049, P31053, Q61470 | |
| Cd37 | |
| CD37 Fusion Protein Ag15384 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction