Learn More
CD370 (Clec9A) Rat anti-Mouse, Brilliant Violet 605, Clone: 10B4 (also known as 24/04-10B4), BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 744511
Description
The 10B4 monoclonal antibody specifically binds to mouse Clec9A. Mouse Clec9A (C-type lectin domain family member 9A) is also known as DNGR1 (Dendritic cell natural killer lectin group receptor 1). It is a type II membrane protein with a single extracellular C-type lectin domain. Clec9A is a dendritic cell subtype-restricted C-type lectin-like receptor. Clec9A is selectively expressed on plasmacytoid dendritic cells and CD8+ myeloid dendritic cells. Clec9A reportedly serves as a receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens in a Syk-dependent manner.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407nm and Em Max of 602nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser. BV605 is a tandem fluorochrome that combines BD Horizon BV421 and Cy™3.5. These conjugates are very bright, exhibiting similar brightness to equivalent PE conjugates. Due to the excitation of Cy3.5 by the green (532nm) and yellow-green (561nm) lasers, there will be significant spillover of the BD Horizon BV605 signal into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers.
Specifications
| CD370 (Clec9A) | |
| 10B4 (also known as 24/04-10B4) | |
| Brilliant Violet 605 | |
| CD370, CLC9A, Clec9a, DNGR-1, C-type lectin domain family 9 member A | |
| Mouse Clec9A Peptide | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.