missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (AT13/5), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81055APC
This item is not returnable.
View return policy
Description
CD38 Monoclonal specifically detects CD38 in Human samples. It is validated for Flow Cytometry.
Specifications
| CD38 | |
| Monoclonal | |
| APC | |
| PBS | |
| CD38 | |
| Namalwa cells emulsified in CFA. | |
| 0.1 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
| 952 | |
| Human | |
| Purified |
| Flow Cytometry | |
| AT13/5 | |
| Flow Cytometry | |
| ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, cADPr hydrolase 1, CD38 antigen, CD38 antigen (p45), CD38 molecule, Cyclic ADP-ribose hydrolase 1, EC 3.2.2.5, NAD(+) nucleosidase, T10 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Binds specifically to human CD38, an approximately 45 kDa type II transmembrane protein, expressed on essentially all pre-B lymphocytes, plasma cells, and thymocytes. Also present on activated T lymphocytes, natural killer (NK) lymphocytes, myeloblasts, and erythroblasts. Bimodally expressed during B cell development, modulating from high in immature cells to low in intermediate ones and back to high on mature B cells. This antibody competes with clone HB7 (Ab00128) (Ellis 1995) which has been shon to bind to an epitope between amino acids 273-285). | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction