missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (CD38/8114R), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21023F
This item is not returnable.
View return policy
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| CD38 | |
| Monoclonal | |
| FITC | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| Recombinant fragment (around aa 200-300) of human CD38 (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
| 952 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| CD38/8114R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction