missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Antibody (HB7), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52696MFV450
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow Cytometry
Spécification
| CD38 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| HB7 | |
| Flow Cytometry | |
| 2'-phospho-ADP-ribosyl cyclase, 2'-phospho-ADP-ribosyl cyclase/2'-phospho-cyclic-ADP-ribose transferase (EC:2.4.99.20), 2'-phospho-cyclic-ADP-ribose transferase, ADPRC 1, ADPRC1, ADP-ribosyl cyclase 1, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase, ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase 1, cADPr hydrolase 1, CD_antigen: CD38, CD38, CD38 antigen, CD38 antigen (p45), CD38 molecule, cluster of differentiation 38, Cyclic ADP-ribose hydrolase 1, EC 2.4.99.20, EC 3.2.2.6, EC:3.2.2.6, NAD(+) nucleosidase, T10 | |
| This CD38 antibody (HB7) was developed against viable cells (1 x 10^7) of the human cell line, BJAB (Human Burkitt lymphoma B cell line), which were injected intraperitoneally into 8-week-old BALB/c mice. | |
| 0.1 mL | |
| Apoptosis, B Cell Development and Differentiation Markers, Cancer, Cell Biology, Hematopoietic Stem Cell Markers, Immunology, Lipid and Metabolism, Neuroscience, Stem Cell Markers | |
| 952 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu