missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD38 Mouse anti-Human, APC-R700, Clone: HIT2, BD Horizon™
Mouse Monoclonal Antibody
Brand: BD Biosciences 564980
This item is not returnable.
View return policy
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow cytometry.
Specifications
| CD38 | |
| Monoclonal | |
| APC-R700 | |
| Aqueous buffered solution containing BSA, protein stabilizer, glycerol and ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon APC-R700 under optimum conditions and unconjugated antibody and free BD Horizon APC-R700 were removed. | |
| IgG1 κ |
| Flow Cytometry | |
| HIT2 | |
| Vol. Per test: 5μL | |
| T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; gp45 | |
| 25 Tests | |
| Cell Biology | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Reseach Use Only.
Spot an opportunity for improvement?Share a Content Correction