Learn More
CD39/ENTPD1 Antibody (12D12), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26601-100ul
 View more versions of this product
Product Code. 30043025
  • € 533.00 / 100µL
Description
CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human samples. It is validated for Flow Cytometry, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD39/ENTPD1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Flow Cytometry, ELISA
12D12
Flow Cytometry 1:50-1:200, ELISA
ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
A synthesized peptide derived from Human CD39/ENTPD1 [UniProt P49961]
100 μL
Lipid and Metabolism, Myeloid derived Suppressor Cell
953
Store at -20 to -70C. Avoid freeze-thaw cycles.