missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39/ENTPD1 Antibody (17B5F11), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-25226AF700
This item is not returnable.
View return policy
Description
CD39/ENTPD1 Monoclonal specifically detects CD39/ENTPD1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CD39/ENTPD1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This product reacts with Human, Mouse and Rat. | |
| Store at 4C in the dark. | |
| IgG2b λ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 17B5F11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ENTPD1 | |
| Amino acids 102-208 of human CD39 were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 953 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction