Learn More
CD39/ENTPD1 Antibody (AC2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50387
This item is not returnable.
View return policy
Description
CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
Specifications
| CD39/ENTPD1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.2 μg/mL, Flow Cytometry 1 μg/505 cells, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
| ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1 | |
| EBV-transformed human B lymphoblastoid cell line | |
| 0.1 mL | |
| Lipid and Metabolism, Myeloid derived Suppressor Cell | |
| 953 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF | |
| AC2 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |