Learn More
CD39/ENTPD1 Antibody (AC2), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50387
 View more versions of this product
Product Code. 18685448
  • € 381.00 / 0.10mL
Description
CD39/ENTPD1 Monoclonal antibody specifically detects CD39/ENTPD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD39/ENTPD1
Monoclonal
1 mg/mL
Western Blot 0.2 μg/mL, Flow Cytometry 1 μg/505 cells, Immunocytochemistry/ Immunofluorescence 1 - 2 μg/mL, Immunoprecipitation 1:10 - 1:500, CyTOF-ready
ATPDase, CD39 antigen, CD39EC 3.6.1.5, DKFZp686D194, DKFZp686I093, EC 3.6.1, Ecto-apyrase, Ecto-ATP diphosphohydrolase 1, Ecto-ATPase 1, Ecto-ATPDase 1, ectonucleoside triphosphate diphosphohydrolase 1, FLJ40921, FLJ40959, Lymphoid cell activation antigen, NTPDase 1, NTPDase-1
EBV-transformed human B lymphoblastoid cell line
0.1 mL
Lipid and Metabolism, Myeloid derived Suppressor Cell
953
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, CyTOF
AC2
Unconjugated
PBS
Mouse
Protein A purified