missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD39L4/ENTPD5 Antibody (064), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89830MFV450
This item is not returnable.
View return policy
Description
CD39L4/ENTPD5 Monoclonal antibody specifically detects CD39L4/ENTPD5 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| CD39L4/ENTPD5 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 064 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| CD39 antigen-like 4, CD39L4NTPDase 5, CD39-like 4, EC 3.6.1, EC 3.6.1.42, EC 3.6.1.6, ectonucleoside triphosphate diphosphohydrolase 5, ER-UDPase, GDPase ENTPD5, Guanosine-diphosphatase ENTPD5, MGC163357, MGC163359, NTPDase-5, Nucleoside diphosphatase, PCPH, Pcph proto-oncogene protein, proto-oncogene CPH, UDPase ENTPD5, Uridine-diphosphatase ENTPD5 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CD39L4/ENTPD5 (Uniprot#: O75356; Met1-His428). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 957 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction