missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3e Armenian Hamster, FITC, Clone: 145-2C11, enQuireBio™
Armenian Hamster Monoclonal Antibody
€ 85.85 - € 225.00
Specifications
Antigen | CD3e |
---|---|
Clone | 145-2C11 |
Concentration | 0.5mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941638
|
EnquireBio
QAB2-F-25UG |
25 μg |
€ 85.85
25µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931638
|
EnquireBio
QAB2-F-100UG |
100 μg |
€ 98.45
100µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15951638
|
EnquireBio
QAB2-F-500UG |
500 μg |
€ 225.00
500µg |
Estimated Shipment: 28-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD3e | |
0.5mg/mL | |
Monoclonal | |
Armenian Hamster | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
12501 | |
IgG | |
Protein A/G Purified |
145-2C11 | |
Flow Cytometry | |
FITC | |
RUO | |
XP_006510029 | |
Cd3e | |
Primary | |
Store at 2-4°C. |