missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD3e/CD3g Recombinant Rabbit Monoclonal Antibody (4F5G4)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA542744

Product Code. 17780015

  • € 528.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The target is usually found in the following locations: Cell membrane, Single-pass type I membrane protein, Membrane, Single-pass type I membrane protein. Immunogen sequence: MQSGTHWRVL GLCLLSVGVW GQDGNEEMGG ITQTPYKVSI SGTTVILTCP QYPGSEILWQ HNDKNIGGDE DDKNIGSDED HLSLKEFSEL EQSGYYVCYP RGSKPEDANF YLYLRARVCE NCMEMDVMSV ATIVIVDICI TGGLLLLVYY WSKNRKAKAK PVTRGAGAGG RQRGQNKERP PPVPNPDYEP IRKGQRDLYS GLNQRRI(CD 3E)&MEQGKG LAVLILAIIL LQGTLAQSIK GNHLVKVYDY QEDGSVLLTC DAEAKNITWF KDGKMIGFLT EDKKKWNLGS NAKDPRGMYQ CKGSQNKSKP LQVYYRMCQN CIELNAATIS GFLFAEIVSI FVLAVGVYFI AGQDGVRQSR ASDKQTLLPN DQLYQPLKDR EDDQYSHLQG NQLRRN(CD3 G).

The CD3 subunit complex which is crucial in transducing antigen-recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR complex. T cell activation through the antigen receptor (TCR) involves the cytoplasmic tails of the CD3 subunits CD3 gamma, CD3 delta, CD3 epsilon and CD3 zeta. These CD3 subunits are structurally related members of the immunoglobulins super family encoded by closely linked genes on human chromosome 11. The CD3 components have long cytoplasmic tails that associate with cytoplasmic signal transduction molecules and this association is mediated at least in part by a double tyrosine-based motif present in a single copy in the CD3 subunits. CD3 may play a role in TCR-induced growth arrest, cell survival and proliferation. The CD3 antigen is present on 68-82% of normal peripheral blood lymphocytes, 65-85% of thymocytes and Purkinje cells in the cerebellum. It is never expressed on B or NK cells. Decreased percentages of T lymphocytes may be observed in some autoimmune diseases. The genes encoding the CD3 epsilon, gamma and delta polypeptides are located on chromosome 11. Defects in the CD3 gene are associated with CD3 immunodeficiency.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD3e/CD3g
Recombinant Monoclonal
0.94 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
P07766, P09693
Rabbit
Affinity Chromatography
RUO
916, 917
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Flow Cytometry, Western Blot
4F5G4
Unconjugated
CD3G
CD3E, CD3G
Recombinant fusion protein containing a sequence corresponding to amino acids 1-207(CD3E)& 1-182(CD3G) of human CD3 epsilon&CD3 gamma Heterodimer.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.