missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMBRD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148382
This item is not returnable.
View return policy
Description
LMBRD2 Polyclonal Antibody for ICC/IF, IHC (P)
Specifications
| LMBRD2 | |
| Polyclonal | |
| Unconjugated | |
| LMBRD2 | |
| G-protein coupled receptor-associated protein LMBRD2; LMBR1 domain containing 2; LMBR1 domain-containing protein 2; LMBRD2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 92255 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q68DH5 | |
| LMBRD2 | |
| E. coli-derived Human LMBRD2 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction