missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3E, Mouse anti-Human, Clone: CL1497, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15531.100uL
This item is not returnable.
View return policy
Description
Sequence: NEEMGGITQTPYKVSISGTTVILTCPQYPGSEILWQHNDKNIGGDEDDKNIGSDEDHLSLKEFSELEQSGYYVCYPRGSKPEDANFYLYLRARVCENCMEMDSpecifications
| CD3E | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
| FLJ18683/T3E/TCRE | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| CL1497 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000 - 1:2500) Western Blot (1:500 - 1:1000) The optimal working dilution should be determined by the end user. | |
| CD3e molecule, epsilon (CD3-TCR complex) | |
| CD3E | |
| Recombinant protein corresponding to amino acids 25-126 of human CD3E. | |
| RUO | |
| 916 | |
| Store at 4°C for short term storage. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction