missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD4, anti-Human, Clone: YNB46.1.8 (Campath-9, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00155.200ug
This item is not returnable.
View return policy
Specifications
| CD4 | |
| Monoclonal | |
| Unconjugated | |
| In PBS, pH 7.2 (0.02% Proclin 300). | |
| CD4mut | |
| Original antibody is raised against rat T-cell line (NB2) transfected with the human CD4. | |
| RUO | |
| 920 | |
| Human | |
| Antibody | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry | |
| YNB46.1.8 (Campath-9 | |
| Flow Cytometry (1 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD4 molecule | |
| CD4 | |
| 200 μg | |
| Primary | |
| This antibody recognizes 55kDa human CD4 cell surface antigen, expressed by helper T lymphocytes and by monocytes. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction